Learn More
Biolegend APC anti-human CD303 BDCA-2
Catalog No. 502722190
Encompass_Preferred
APC anti-human CD303 (BDCA-2) [201A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2190 Supplier Biolegend Supplier No. 354205
APC anti-human CD303 (BDCA-2) [201A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests