Learn More
Biolegend APC anti-human CD307e (FcRL5)
Supplier:  Biolegend 340306
Encompass_Preferred
APC anti-human CD307e (FcRL5) [509f6]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1286509