Learn More
Biolegend APC anti-human CD338 (ABCG2)
Supplier:  Biolegend 332020
Encompass_Preferred
APC anti-human CD338 (ABCG2) [5D3]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-167-704