Learn More
Cytek Biosciences Inc Antibodies APC Anti-Human CD34 (4H11) ..... Cat. No. 20-0349-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 200349T025
Encompass_Preferred
Antibodies APC Anti-Human CD34 (4H11) ..... Cat. No. 20-0349-T025 ..... Size 25 testsThe 4H11 monoclonal antibody reacts with human CD34, a single pass transmembrane protein with a molecular weight of 105-120 kDa. CD34 is heavily glycosylated with an N-terminal mucin domain. Although the function of CD34 is not defined, the intracellular domain of CD34 is a target for phosphorylation by activated protein kinase C, suggesting a possible role in signal transduction. CD34 is expressed on hematopoietic progenitor cells, some populations of mesenchymal stem cells, and vascular endothelium. The epitope of CD34 recognized by the 4H11 antibody is assigned to the class III epitopes. Additionally, CD34 may also function as an adhesion molecule with a role in mediating attachment of stem cells to bone marrow extracellular matrix, stromal cells, or other bone marrow components.
Catalog No. 50-210-2732