Learn More
Biolegend APC anti-human CD360 (IL-21R)
Supplier:  Biolegend 347808
Encompass_Preferred
APC anti-human CD360 (IL-21R) [2G1-K12]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1201196