Learn More
Biolegend APC anti-human CD369 (Dectin-1/CLEC7A)
Supplier:  Biolegend 355406
Encompass_Preferred
APC anti-human CD369 (Dectin-1/CLEC7A) [15E2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1885447