Learn More
Biolegend APC anti-human CD38
Supplier:  Biolegend 397112
Encompass_Preferred
APC anti-human CD38 [S17015A]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-9285