Learn More
Biolegend APC anti-human CD71 [CY1G4]; Isotype: Mouse IgG2a, ; Reactivity: Human; Apps: FC; Size: 25 tests
Supplier:  Biolegend 334107
Encompass_Preferred
APC anti-human CD71 [CY1G4]; Isotype: Mouse IgG2a, ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-167-761