Learn More
Biolegend APC anti-human CD96 (TACTILE)
Supplier:  Biolegend 338410
Encompass_Preferred
APC anti-human CD96 (TACTILE) [NK92.39]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1112277