Learn More
Biolegend APC anti-human CD99
Supplier:  Biolegend 398204
Encompass_Preferred
APC anti-human CD99 [hec2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-9352