Learn More
Biolegend APC anti-human GM-CSF
Supplier:  Biolegend 502310
Encompass_Preferred
APC anti-human GM-CSF [BVD2-21C11]; Isotype: Rat IgG2a, κ; Reactivity: Human, Cross-Reactivity: Rhesus, Cynomolgus; Apps: ICFC; Size: 100 tests
Catalog No. NC1340839