Learn More
Biolegend APC anti-human HLA-DR
Supplier:  Biolegend 361609
Encompass_Preferred
APC anti-human HLA-DR [Tü36]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Baboon, Cynomolgus, Cat (Feline), Dog (Canine), Cattle (Bovine, Cow); Apps: FC; Size: 25 tests
Catalog No. NC1245667