Learn More
Biolegend APC anti-human LAP (TGF-β1)
Supplier:  Biolegend 300006
Encompass_Preferred
APC anti-human LAP (TGF-β1) [S20006A]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-213-1854