Learn More
Biolegend APC anti-human PSMA (FOLH1)
Supplier:  Biolegend 342508
Encompass_Preferred
APC anti-human PSMA (FOLH1) [LNI-17]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1913424