Learn More
Biolegend APC anti-human/rat CD51 (Integrin αvβ5) [P1F6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Rat; Apps: FC; Size: 100 tests
Supplier:  Biolegend 920012
Encompass_Preferred
APC anti-human/rat CD51 (Integrin αvβ5) [P1F6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Rat; Apps: FC; Size: 100 tests
Catalog No. 50-237-5017