Learn More
Biolegend APC anti-mouse IL-23R
Supplier:  Biolegend 150906
Encompass_Preferred
APC anti-mouse IL-23R [12B2B64]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1547385