Learn More
Biolegend APC anti-mouse β2-microglobulin
Supplier:  Biolegend 154505
Encompass_Preferred
APC anti-mouse β2-microglobulin [A16041A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC1509808