Learn More
Biolegend APC anti-mouse CD100 (SEMA4D)
Supplier:  Biolegend 147605
Encompass_Preferred
APC anti-mouse CD100 (SEMA4D) [BMA-12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-1755