Learn More
Biolegend APC anti-mouse CD105
Catalog No. 502722738
Encompass_Preferred
APC anti-mouse CD105 [MJ7/18]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Catalog No. 50-272-2738 Supplier Biolegend Supplier No. 120414
APC anti-mouse CD105 [MJ7/18]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg