Learn More
Cytek Biosciences Inc Antibodies APC Anti-Mouse CD134 (OX40) (OX-86) ..... Cat. No. 20-1341-U100 ..... Size 100 µg
Supplier:  Cytek Biosciences Inc 201341U100
Encompass_Preferred
Antibodies APC Anti-Mouse CD134 (OX40) (OX-86) ..... Cat. No. 20-1341-U100 ..... Size 100 µgThe OX-86 monoclonal antibody reacts with mouse CD134, a 50kD type 1 integral membrane glycoprotein and a member of the TNF-receptor superfamily. CD134 is also known as OX40 and is expressed on activated CD4 and CD8 T cells. CD134 is the receptor for CD252 (OX40 ligand) and the interaction of this receptor-ligand pair is implicated in T-dependent humoral responses as well as numerous aspects of T cell expansion, survival and memory. Additionally, CD134 seems to play a significant role in the pathogenesis of some autoimmune diseases.
Catalog No. 50-210-2755