Learn More
Biolegend APC anti-mouse CD152
Supplier:  Biolegend 106310
Encompass_Preferred
APC anti-mouse CD152 [UC10-4B9]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC0364939