Learn More
Biolegend APC anti-mouse CD154
Supplier:  Biolegend 106509
Encompass_Preferred
APC anti-mouse CD154 [MR1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC1366456