Learn More
Biolegend APC anti-mouse CD273 (B7-DC, PD-L2)
Supplier:  Biolegend 107210
Encompass_Preferred
APC anti-mouse CD273 (B7-DC, PD-L2) [TY25]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1047861