Learn More
Cytek Biosciences Inc Antibodies APC Anti-Mouse CD366 (TIM-3) (RMT3-23) ..... Cat. No. 20-5870-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 205870U025
Encompass_Preferred
Antibodies APC Anti-Mouse CD366 (TIM-3) (RMT3-23) ..... Cat. No. 20-5870-U025 ..... Size 25 µgThe RMT3-23 antibody reacts with mouse CD366 which is also known as Tim-3 (T cell immunoglobulin and mucin domain containing protein-3). Tim-3 is expressed on activated monocytes, macrophages, dendritic cells, and mast cells. Additionally, Tim-3 is found on Th1 lymphocytes and functions as an inhibitory receptor involved in the maintenance of immune homeostasis and self tolerance.
Catalog No. 50-210-2770