Learn More
Biolegend APC anti-mouse CD370 (CLEC9A, DNGR1)
Supplier:  Biolegend 143505
Encompass_Preferred
APC anti-mouse CD370 (CLEC9A, DNGR1) [7H11]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC1543781