Learn More
Cytek Biosciences Inc Antibodies APC Anti-Mouse CD49b (HMa2) ..... Cat. No. 20-0491-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 200491U025
Encompass_Preferred
Antibodies APC Anti-Mouse CD49b (HMa2) ..... Cat. No. 20-0491-U025 ..... Size 25 µgThe HMa2 antibody reacts with mouse CD49b, a 150 kDa glycoprotein member of the integrin family. CD49b, also known as integrin alpha2, associates non-covalently with CD29 (integrin beta1) to form the VLA-2 heterodimer. VLA-2 integrin s function as cellular receptors for collagen and laminin. CD49b is expressed by platelets, intestinal epithelial cells, NK cells, epithelial cells, and a subset of CD4+ T cells. CD49b plays an essential role in mediating the adhesion of cells to the extracellular matrix.
Catalog No. 50-210-2741