Learn More
Biolegend APC anti-mouse CD5
Supplier:  Biolegend 100625
Encompass_Preferred
APC anti-mouse CD5 [53-7.3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC1926918