Learn More
Biolegend APC anti-mouse CD69 Recombinan
Catalog No. 5000064557
Encompass_Preferred
APC anti-mouse CD69 Recombinant [QA17A41] Isotype Rat IgG1 Reactivity Mouse Apps FC Size 100 g
Catalog No. 50-000-64557 Supplier Biolegend Supplier No. 164206
APC anti-mouse CD69 Recombinant [QA17A41] Isotype Rat IgG1 Reactivity Mouse Apps FC Size 100 g