Learn More
Biolegend APC anti-mouse/human CD324 (E-Cadherin)
Supplier:  Biolegend 147311
Encompass_Preferred
APC anti-mouse/human CD324 (E-Cadherin) [DECMA-1]; Isotype: Rat IgG1, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Dog, Swine; Apps: FC; Size: 25 ug
Catalog No. 50-207-1741