Learn More
Biolegend APC anti-mouse/human CD44 Recombinant
Supplier:  Biolegend 163603
Encompass_Preferred
APC anti-mouse/human CD44 Recombinant [QA19A43]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human; Apps: FC; Size: 25 ug
Catalog No. 50-213-1838