Learn More
Biolegend APC anti-mouse IL-33Rα (IL1RL1, ST2)
Supplier:  Biolegend 145305
Encompass_Preferred
APC anti-mouse IL-33Rα (IL1RL1, ST2) [DIH9]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. NC0527471