Learn More
Biolegend APC anti-mouse IL-33Rα (ST2)
Supplier:  Biolegend 146606
Encompass_Preferred
APC anti-mouse IL-33Rα (ST2) [DIH4]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-1709