Learn More
Cytek Biosciences Inc Antibodies APC Anti-Mouse LPAM-1 (DATK32) ..... Cat. No. 20-5887-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 205887U025
Encompass_Preferred
Antibodies APC Anti-Mouse LPAM-1 (DATK32) ..... Cat. No. 20-5887-U025 ..... Size 25 µgThe DATK32 antibody is specific for a combinatorial epitope of the mouse integrin alpha 4/beta 7 (LPAM-1) heterodimer. LPAM-1 is expressed on most mature lymphocytes and sub-populations of thymocytes and bone marrow cells. It is involved in lymphocyte adhesion and migration and is a receptor for several extracellular matrix proteins including VCAM-1, fibronectin, and MAdCAM-1.DATK32 is reported to induce LPAM-1 dependent lymphocyte aggregation and block adhesion to VCAM-1, fibronectin, and MAdCAM-1.
Catalog No. 50-210-2772