Learn More
Biolegend APC anti-mouse/rat CD62P (P-selectin)
Supplier:  Biolegend 148304
Encompass_Preferred
APC anti-mouse/rat CD62P (P-selectin) [RMP-1]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse, Rat; Apps: FC; Size: 100 ug
Catalog No. NC1363351