Learn More
Biolegend APC anti-mouse/rat CD81
Supplier:  Biolegend 104910
Encompass_Preferred
APC anti-mouse/rat CD81 [Eat-2]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: FC; Size: 100 ug
Catalog No. NC1486588