Learn More
Biolegend APC anti-mouse/rat TNF-alpha
Catalog No. 502721885
Encompass_Preferred
APC anti-mouse/rat TNF-α [TN3-19.12]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: ICFC; Size: 100 μg
Catalog No. 50-272-1885 Supplier Biolegend Supplier No. 506108
APC anti-mouse/rat TNF-α [TN3-19.12]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: ICFC; Size: 100 μg