Learn More
Biolegend APC anti-mouse B7-H4 (B7S1, B7X)
Supplier:  Biolegend 139408
Encompass_Preferred
APC anti-mouse B7-H4 (B7S1, B7X) [HMH4-5G1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-103-0561