Learn More
Biolegend APC anti-mouse CD11a
Supplier:  Biolegend 101120
Encompass_Preferred
APC anti-mouse CD11a [M17/4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-206-9540