Learn More
Biolegend APC anti-mouse CD11c
Supplier:  Biolegend 117309
Encompass_Preferred
APC anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-163-705