Learn More
Biolegend APC anti-mouse CD134 (OX-40)
Supplier:  Biolegend 119413
Encompass_Preferred
APC anti-mouse CD134 (OX-40) [OX-86]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0351