Learn More
Biolegend APC anti-mouse CD150 (SLAM)
Supplier:  Biolegend 115910
Encompass_Preferred
APC anti-mouse CD150 (SLAM) [TC15-12F12.2]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-584