Learn More
Biolegend APC anti-mouse CD2
Supplier:  Biolegend 100112
Encompass_Preferred
APC anti-mouse CD2 [RM2-5]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-206-9418