Learn More
Biolegend APC anti-mouse CD200R3
Supplier:  Biolegend 142208
Encompass_Preferred
APC anti-mouse CD200R3 [Ba13]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-605-066