Learn More
Biolegend APC anti-mouse CD202b (Tie-2, CD202)
Supplier:  Biolegend 124009
Encompass_Preferred
APC anti-mouse CD202b (Tie-2, CD202) [TEK4]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-156