Learn More
Biolegend APC anti-mouse CD206 (MMR)
Supplier:  Biolegend 141708
Encompass_Preferred
APC anti-mouse CD206 (MMR) [C068C2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, FC; Size: 100 ug
Catalog No. 50-165-219