Learn More
Biolegend APC anti-mouse CD226 (DNAM-1)
Supplier:  Biolegend 128809
Encompass_Preferred
APC anti-mouse CD226 (DNAM-1) [10E5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0877