Learn More
Biolegend APC anti-mouse CD278 (ICOS)
Supplier:  Biolegend 117420
Encompass_Preferred
APC anti-mouse CD278 (ICOS) [7E.17G9]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-0310