Learn More
Biolegend APC anti-mouse CD28
Supplier:  Biolegend 102109
Encompass_Preferred
APC anti-mouse CD28 [37.51]; Isotype: Syrian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-643