Learn More
Biolegend APC anti-mouse CD3
Supplier:  Biolegend 100236
Encompass_Preferred
APC anti-mouse CD3 [17A2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-402-914