Learn More
Biolegend APC anti-mouse CD335 (NKp46)
Supplier:  Biolegend 137608
Encompass_Preferred
APC anti-mouse CD335 (NKp46) [29A1.4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-164-983